siRNA Details
Gene Symbol: MYCBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012333.4 | NP_036465.2 | 5 | 357 | ||
NR_037632.1 | 4 | 298 | ||||
GenBank mRNA | AB007191.2 | - | 5 | 322 | ||
AB451287.1 | - | 5 | 284 | |||
AB451419.1 | - | 5 | 284 | |||
AK311886.1 | - | 5 | 347 | |||
BC008686.1 | - | 5 | 322 | 3870100 | ||
BQ437941.1 | - | 4 | 262 | |||
BT007109.1 | - | 5 | 284 | |||
BU601195.1 | - | 5 | 359 | |||
CR456936.1 | - | 5 | 284 | |||
D50692.1 | - | 5 | 330 |
Gene Symbol: GJA9-MYCBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037633.1 | 6 | 3063 | |||
NR_037634.1 | 7 | 2195 | ||||
NR_037635.1 | 5 | 464 | ||||
NR_037636.1 | 4 | 391 | ||||
NR_037637.1 | 3 | 332 | ||||
GenBank mRNA | AK123051.1 | - | 7 | 2189 | ||
BC039090.1 | - | 5 | 1084 | |||
BC051675.1 | - | 6 | 3035 | |||
FY210901.1 | - | 3 | 308 | |||
FY211253.1 | - | 4 | 367 | |||
FY211295.1 | - | 5 | 440 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
