siRNA Details
Gene Symbol: MYCBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012333.4 | NP_036465.2 | 5 | 506 | ||
NR_037632.1 | 4 | 447 | ||||
GenBank mRNA | AB007191.2 | - | 5 | 471 | ||
BC008686.1 | - | 5 | 471 | 3870100 | ||
BQ437941.1 | - | 4 | 411 | |||
BU601195.1 | - | 5 | 508 | |||
D50692.1 | - | 5 | 479 |
Gene Symbol: GJA9-MYCBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037633.1 | 6 | 3212 | |||
NR_037634.1 | 7 | 2344 | ||||
NR_037635.1 | 5 | 613 | ||||
NR_037636.1 | 4 | 540 | ||||
NR_037637.1 | 3 | 481 | ||||
GenBank mRNA | AK123051.1 | - | 7 | 2338 | ||
BC039090.1 | - | 5 | 1233 | |||
BC051675.1 | - | 6 | 3184 | |||
FY210901.1 | - | 3 | 457 | |||
FY211253.1 | - | 4 | 516 | |||
FY211295.1 | - | 5 | 589 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
