siRNA Details
Gene Symbol: ZNF708 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297560.1 | NP_001284489.1 | 4 | 1234 | ||
NM_001297561.1 | NP_001284490.1 | 3 | 1025 | |||
NM_021269.2 | NP_067092.2 | 4 | 1150 | |||
XM_017027201.1 | XP_016882690.1 | 4 | 1052 | |||
XM_017027202.1 | XP_016882691.1 | 4 | 1175 | |||
XM_017027203.1 | XP_016882692.1 | 2 | 955 | |||
XM_017027204.1 | XP_016882693.1 | 4 | 967 | |||
GenBank mRNA | AK128184.1 | - | 4 | 1109 | ||
AK131527.1 | - | 4 | 1150 | |||
BX647526.1 | - | 4 | 1220 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
