siRNA Details
Gene Symbol: FAM19A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001082967.2 | NP_001076436.1 | 4 | 1290 | ||
NM_015381.6 | NP_056196.2 | 4 | 1311 | |||
GenBank mRNA | AF131851.1 | - | 4 | 1256 | ||
AK123443.1 | - | 5 | 1250 | |||
AK125256.1 | - | 4 | 1311 | |||
AY325118.1 | - | 4 | 1285 | |||
AY358847.1 | - | 5 | 1186 | |||
BC039396.1 | - | 4 | 1270 | 5299791 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
