siRNA Details
Gene Symbol: TECPR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172631.2 | NP_001166102.1 | 3 | 512 | ||
NM_014844.4 | NP_055659.2 | 3 | 512 | |||
GenBank mRNA | AB002327.1 | - | 3 | 461 | ||
AK127721.1 | - | 3 | 945 | |||
AK296395.1 | - | 3 | 363 | |||
BC136647.1 | - | 2 | 309 | 9020637 | ||
BC142667.1 | - | 3 | 345 | 40148157 | ||
BC142715.1 | - | 3 | 345 | 40148843 | ||
BC151230.1 | - | 3 | 485 | 8860116 | ||
BP365224.1 | - | 3 | 514 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
