siRNA Details
Gene Symbol: ZNF28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006969.3 | NP_008900.3 | 4 | 3616 | ||
NR_036599.1 | 5 | 3734 | ||||
NR_036600.1 | 3 | 3489 | ||||
XM_006723354.3 | XP_006723417.1 | 4 | 3591 | |||
XM_011527262.2 | XP_011525564.1 | 4 | 3646 | |||
GenBank mRNA | AK091553.1 | - | 1 | 1521 | ||
AK092385.1 | - | 1 | 2423 | |||
AK131368.1 | - | 2 | 4211 | |||
AK131574.1 | - | 1 | 3001 | |||
AK304704.1 | - | 4 | 3577 | |||
BC140720.1 | - | 3 | 3515 | 9021588 | ||
BX640767.1 | - | 1 | 2644 | |||
CR933598.1 | - | 4 | 3576 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
