siRNA Details
Gene Symbol: ZNF28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006969.3 | NP_008900.3 | 4 | 4158 | ||
NR_036599.1 | 5 | 4276 | ||||
NR_036600.1 | 3 | 4031 | ||||
XM_006723354.3 | XP_006723417.1 | 4 | 4133 | |||
XM_011527262.2 | XP_011525564.1 | 4 | 4188 | |||
GenBank mRNA | AK091553.1 | - | 1 | 2063 | ||
AK092385.1 | - | 1 | 2965 | |||
AK131368.1 | - | 2 | 4753 | |||
AK131574.1 | - | 1 | 3543 | |||
AK304704.1 | - | 4 | 4120 | |||
BX640767.1 | - | 1 | 3186 | |||
CR933598.1 | - | 4 | 4118 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
