siRNA Details
Gene Symbol: ZNF860 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001137674.2 | NP_001131146.2 | 2 | 2923 | ||
XM_017006299.1 | XP_016861788.1 | 2 | 4006 | |||
GenBank mRNA | AK294003.1 | - | 2 | 2923 |
Gene Symbol: ZNF28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006969.3 | NP_008900.3 | 4 | 4335 | ||
NR_036599.1 | 5 | 4453 | ||||
NR_036600.1 | 3 | 4208 | ||||
XM_006723354.3 | XP_006723417.1 | 4 | 4310 | |||
XM_011527262.2 | XP_011525564.1 | 4 | 4365 | |||
GenBank mRNA | AK091553.1 | - | 1 | 2240 | ||
AK092385.1 | - | 1 | 3142 | |||
AK131368.1 | - | 2 | 4930 | |||
AK131574.1 | - | 1 | 3720 | |||
AK304704.1 | - | 4 | 4297 | |||
BX640767.1 | - | 1 | 3363 | |||
CR933598.1 | - | 4 | 4295 |
Gene Symbol: ZNF761 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008401.3 | NP_001008401.3 | 6 | 3950 | ||
NM_001289951.1 | NP_001276880.1 | 5 | 3731 | |||
NM_001289952.1 | NP_001276881.1 | 5 | 3799 | |||
GenBank mRNA | AB107355.2 | - | 5 | 3800 | ||
AK092652.1 | - | 1 | 2284 | |||
AK131281.1 | - | 2 | 3420 |
Gene Symbol: ZNF813 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004301.3 | NP_001004301.2 | 4 | 3301 | ||
GenBank mRNA | AK091460.1 | - | 1 | 2451 | ||
AK131422.1 | - | 1 | 2684 |
Gene Symbol: ZNF888 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005259451.4 | XP_005259508.1 | 5 | 3582 | ||
XM_017026798.1 | XP_016882287.1 | 5 | 3743 | |||
XM_017026799.1 | XP_016882288.1 | 5 | 3628 | |||
XM_017026800.1 | XP_016882289.1 | 3 | 3359 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
