siRNA Details
Gene Symbol: VPS29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282150.1 | NP_001269079.1 | 5 | 494 | ||
NM_016226.4 | NP_057310.1 | 3 | 339 | |||
NM_057180.2 | NP_476528.1 | 4 | 351 | |||
NR_104099.1 | 5 | 391 | ||||
NR_104100.1 | 4 | 379 | ||||
NR_104101.1 | 4 | 429 | ||||
NR_104102.1 | 3 | 230 | ||||
XM_006719459.3 | XP_006719522.1 | 5 | 400 | |||
XM_006719460.3 | XP_006719523.1 | 6 | 437 | |||
XM_011538484.2 | XP_011536786.1 | 4 | 385 | |||
XM_017019457.1 | XP_016874946.1 | 4 | 410 | |||
XM_017019458.1 | XP_016874947.1 | 3 | 530 | |||
GenBank mRNA | AA148059.1 | - | 3 | 177 | ||
AB578890.1 | - | 2 | 78 | |||
AF168716.1 | - | 4 | 309 | |||
AF175264.1 | - | 3 | 308 | |||
AF193795.1 | - | 3 | 301 | |||
AF201936.1 | - | 4 | 396 | |||
AF201946.1 | - | 3 | 287 | |||
AK000499.1 | - | 2 | 67 | |||
AK129767.1 | - | 3 | 301 | |||
AK130510.1 | - | 1137 | ||||
AL136614.1 | - | 3 | 317 | |||
AL832866.1 | - | 3 | 2332 | |||
BC000880.1 | - | 4 | 309 | 3461977 | ||
BC015095.1 | - | 4 | 318 | |||
BC095446.1 | - | 3 | 310 | 30719438 | ||
BF030144.1 | - | 349 | ||||
BG707325.1 | - | 3 | 334 | |||
BM806646.1 | - | 5 | 447 | |||
BU568689.1 | - | 5 | 352 | |||
CR457182.1 | - | 2 | 262 | |||
CR533468.1 | - | 2 | 250 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
