siRNA Details
Gene Symbol: VPS29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282150.1 | NP_001269079.1 | 6 | 789 | ||
NM_001282151.1 | NP_001269080.1 | 3 | 398 | |||
NM_016226.4 | NP_057310.1 | 4 | 634 | |||
NM_057180.2 | NP_476528.1 | 5 | 646 | |||
NR_104099.1 | 6 | 686 | ||||
NR_104100.1 | 5 | 674 | ||||
NR_104101.1 | 5 | 724 | ||||
NR_104102.1 | 4 | 525 | ||||
XM_006719459.3 | XP_006719522.1 | 6 | 695 | |||
XM_006719460.3 | XP_006719523.1 | 7 | 732 | |||
XM_011538484.2 | XP_011536786.1 | 5 | 680 | |||
XM_017019457.1 | XP_016874946.1 | 5 | 705 | |||
XM_017019458.1 | XP_016874947.1 | 4 | 825 | |||
XM_017019459.1 | XP_016874948.1 | 4 | 487 | |||
GenBank mRNA | AF168716.1 | - | 5 | 604 | ||
AF175264.1 | - | 4 | 603 | |||
AF193795.1 | - | 4 | 596 | |||
AF201936.1 | - | 5 | 691 | |||
AF201946.1 | - | 4 | 582 | |||
AK000499.1 | - | 3 | 362 | |||
AK129767.1 | - | 4 | 596 | |||
AK130510.1 | - | 1432 | ||||
AL136614.1 | - | 4 | 612 | |||
AL832866.1 | - | 4 | 2627 | |||
BC000880.1 | - | 5 | 604 | 3461977 | ||
BC015095.1 | - | 5 | 613 | |||
BC095446.1 | - | 4 | 605 | 30719438 | ||
BG707325.1 | - | 4 | 629 | |||
BM806646.1 | - | 6 | 742 | |||
BU568689.1 | - | 6 | 647 | |||
CB158821.1 | - | 3 | 332 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
