siRNA Details
Gene Symbol: GNL3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184819.1 | NP_001171748.1 | 9 | 913 | ||
NM_019067.5 | NP_061940.1 | 9 | 796 | |||
GenBank mRNA | AK001475.1 | - | 9 | 796 | ||
BC003603.1 | - | 9 | 821 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
