siRNA Details
Gene Symbol: PJA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001032396.2 | NP_001027568.1 | 2 | 1024 | ||
NM_022368.4 | NP_071763.2 | 3 | 786 | |||
NM_145119.3 | NP_660095.1 | 2 | 1350 | |||
XM_005262292.1 | XP_005262349.1 | 2 | 1012 | |||
XM_011531011.2 | XP_011529313.1 | 2 | 1157 | |||
GenBank mRNA | AF262024.1 | - | 1 | 1279 | ||
AF264620.1 | - | 3 | 701 | |||
AK021892.1 | - | 1 | 129 | |||
AK096611.1 | - | 2 | 1349 | |||
AK226171.1 | - | 2 | 931 | |||
AK290543.1 | - | 2 | 1250 | |||
AK293266.1 | - | 3 | 830 | |||
AK294632.1 | - | 3 | 863 | |||
BC039149.1 | - | 1 | 356 | |||
BC048323.1 | - | 1 | 196 | |||
BC058327.1 | - | 1 | 336 | |||
BC075803.1 | - | 2 | 1257 | 3913310 | ||
BC105051.1 | - | 1 | 1039 | 8144054 | ||
BC105053.1 | - | 1 | 1039 | 8144056 | ||
BC113035.1 | - | 1 | 360 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
