siRNA Details
Gene Symbol: PRRC2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004638.3 | NP_004629.3 | 12 | 1655 | ||
NM_080686.2 | NP_542417.2 | 12 | 1618 | |||
XM_017011274.1 | XP_016866763.1 | 11 | 1370 | |||
GenBank mRNA | AB210034.1 | - | 12 | 1604 | ||
AK302756.1 | - | 12 | 1579 | |||
AK309774.1 | - | 12 | 1579 | |||
BC042295.1 | - | 12 | 1581 | 5555847 | ||
BC060668.1 | - | 12 | 1542 | 6471026 | ||
M33509.1 | - | 13 | 1434 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
