siRNA Details
Gene Symbol: PRRC2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004638.3 | NP_004629.3 | 23 | 5643 | ||
NM_080686.2 | NP_542417.2 | 23 | 5606 | |||
XM_017011274.1 | XP_016866763.1 | 22 | 5358 | |||
GenBank mRNA | AB210034.1 | - | 23 | 3720 | ||
AK297210.1 | - | 10 | 3122 | |||
AK298584.1 | - | 7 | 943 | |||
BC030127.1 | - | 7 | 976 | |||
BC042295.1 | - | 23 | 5569 | 5555847 | ||
BC060668.1 | - | 23 | 5530 | 6471026 | ||
CR749245.1 | - | 8 | 2748 | |||
M33509.1 | - | 24 | 5413 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
