siRNA Details
Gene Symbol: PRRC2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004638.3 | NP_004629.3 | 23, 24 | 5667 | ||
NM_080686.2 | NP_542417.2 | 23, 24 | 5630 | |||
XM_017011274.1 | XP_016866763.1 | 22, 23 | 5382 | |||
GenBank mRNA | AB210034.1 | - | 23, 24 | 3744 | ||
AK297210.1 | - | 10, 11 | 3146 | |||
AK298584.1 | - | 7, 8 | 967 | |||
BC030127.1 | - | 7, 8 | 1000 | |||
BC042295.1 | - | 23, 24 | 5593 | 5555847 | ||
BC060668.1 | - | 23, 24 | 5554 | 6471026 | ||
CR749245.1 | - | 8, 9 | 2772 | |||
M33509.1 | - | 24, 25 | 5437 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
