siRNA Details
Gene Symbol: STAG3L5P-PVRIG2P-PILRB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF217981.1 | - | 5 | 1164 | ||
AK074039.1 | - | 5 | 2978 | |||
AK298676.1 | - | 1 | 70 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
