siRNA Details
Gene Symbol: SPRR2G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014291.3 | NP_001014313.1 | 2 | 138 | ||
XM_017002177.1 | XP_016857666.1 | 4 | 515 | |||
XM_017002178.1 | XP_016857667.1 | 2 | 61 | |||
GenBank mRNA | AA490630.1 | - | 1 | 48 | ||
BC130455.1 | - | 2 | 138 | |||
BC133041.1 | - | 2 | 138 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
