siRNA Details
Gene Symbol: SPRR2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003062.1 | 2 | 256 | |||
GenBank mRNA | BC120937.1 | - | 2 | 214 | ||
M21539.1 | - | 2 | 256 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
