siRNA Details
Gene Symbol: MRAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178817.3 | NP_848932.1 | 3 | 276 | ||
NM_206898.1 | NP_996781.1 | 3 | 276 | |||
XM_006724028.3 | XP_006724091.1 | 1 | 206 | |||
XM_017028407.1 | XP_016883896.1 | 1 | 202 | |||
GenBank mRNA | AF454915.1 | - | 3 | 276 | ||
AF483549.1 | - | 1 | 89 | |||
AY079152.1 | - | 1 | 89 | |||
BC062721.1 | - | 1 | 214 | 30327887 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_029844, AY079154, AK003912, BC027543, AK158113, and others.
Gene Ontology Categories:
