siRNA Details
Gene Symbol: MZT2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085365.1 | NP_001078834.1 | 3 | 575 | ||
GenBank mRNA | BC018206.1 | - | 3 | 479 | 3862861 | |
BC063024.1 | - | 3 | 458 | |||
BC104650.1 | - | 2 | 356 | |||
BM554057.1 | - | 3 | 637 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
