siRNA Details
Gene Symbol: MOBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278322.1 | NP_001265251.1 | 3 | 324 | ||
NM_001278323.1 | NP_001265252.1 | 1 | 72 | |||
NM_182935.3 | NP_891980.1 | 3 | 324 | |||
NR_003090.2 | 3 | 324 | ||||
NR_103504.1 | 3 | 324 | ||||
NR_103505.1 | 3 | 324 | ||||
GenBank mRNA | AK127034.1 | - | 3 | 220 | ||
AK290187.1 | - | 3 | 215 | |||
BC022471.1 | - | 3 | 218 | |||
BC075796.1 | - | 3 | 292 | |||
BC119694.1 | - | 2 | 101 | 40116789 | ||
BC119695.1 | - | 2 | 101 | 40116791 | ||
D28113.1 | - | 1 | 76 | |||
D28114.1 | - | 1 | 76 | |||
DA159663.1 | - | 4 | 463 | |||
DA343382.1 | - | 3 | 218 | |||
DA418624.1 | - | 3 | 318 | |||
EB387411.1 | - | 3 | 192 | |||
KU178193.1 | - | 1 | 68 | |||
KU178194.1 | - | 1 | 68 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
