siRNA Details
Gene Symbol: ACOT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037161.1 | NP_001032238.1 | 2 | 824 | ||
XM_017021589.1 | XP_016877078.1 | 2 | 290 | |||
XM_017021590.1 | XP_016877079.1 | 4 | 308 | |||
XM_017021591.1 | XP_016877080.1 | 5 | 791 | |||
XM_017021592.1 | XP_016877081.1 | 2 | 321 | |||
GenBank mRNA | AK300886.1 | - | 2 | 861 | ||
BC127747.1 | - | 2 | 706 | 40127752 | ||
BC127748.1 | - | 2 | 706 | 40127753 | ||
BC132889.1 | - | 2 | 654 | 40146911 | ||
BC132891.1 | - | 2 | 654 | 40146913 | ||
BC143042.1 | - | 2 | 705 | 40127754 | ||
BX161396.1 | - | 2 | 934 | |||
DQ082754.1 | - | 2 | 824 | |||
GU727640.1 | - | 2 | 626 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
