siRNA Details
Gene Symbol: WDFY3-AS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015359.1 | 1 | 202 | |||
GenBank mRNA | AY250185.1 | - | 2 | 555 | ||
BC019264.1 | - | 2 | 108 | |||
BC033127.1 | - | 1 | 183 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
