siRNA Details
Gene Symbol: SNX20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144972.1 | NP_001138444.1 | 2 | 188 | ||
NM_153337.2 | NP_699168.1 | 2 | 188 | |||
NM_182854.3 | NP_878274.1 | 2 | 188 | |||
GenBank mRNA | AA278249.1 | - | 2 | 189 | ||
AK055837.1 | - | 2 | 153 | |||
AK292650.1 | - | 2 | 147 | |||
AK292705.1 | - | 2 | 153 | |||
AY302441.1 | - | 2 | 90 | |||
BC027944.1 | - | 2 | 137 | |||
BC063423.1 | - | 2 | 129 | |||
BC119626.1 | - | 2 | 41 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
