siRNA Details
Gene Symbol: PDXDC2P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003610.1 | 15, 16 | 1455 | |||
GenBank mRNA | AK292860.1 | - | 15, 16 | 1325 | ||
AL833251.1 | - | 15, 16 | 1455 | |||
BC063633.1 | - | 14, 15 | 1327 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
