siRNA Details
Gene Symbol: FAM86B3P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024361.1 | 5 | 444 | |||
NR_024362.1 | 5 | 541 | ||||
NR_024363.1 | 4 | 439 | ||||
GenBank mRNA | AK304567.1 | - | 5 | 513 | ||
AK307256.1 | - | 4 | 409 | |||
BC035792.1 | - | 5 | 419 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
