siRNA Details
Gene Symbol: VCX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013452.2 | NP_038480.2 | 3 | 541 | ||
XM_011545490.2 | XP_011543792.1 | 2 | 433 | |||
GenBank mRNA | AF159129.1 | - | 2 | 348 | ||
AF167081.2 | - | 3 | 541 | |||
BC126903.1 | - | 2 | 438 | 40030361 |
Gene Symbol: VCX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF159127.1 | - | 2 | 347 | ||
AF167079.1 | - | 3 | 629 | |||
BC096715.1 | - | 2 | 347 | 40002645 | ||
BC098101.1 | - | 2 | 347 | 40002641 | ||
BC098113.1 | - | 2 | 347 | 40014419 | ||
BC098163.1 | - | 2 | 347 | 40014421 | ||
BC104153.2 | - | 2 | 347 | 40029400 | ||
BC104167.1 | - | 2 | 438 | 40030354 | ||
BC104168.1 | - | 2 | 438 | 40030355 |
Gene Symbol: VCX3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF159128.1 | - | 2 | 328 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
