siRNA Details
Gene Symbol: IGLL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001178126.1 | NP_001171597.1 | 3 | 837 | ||
NM_001256296.1 | NP_001243225.1 | 2 | 718 | |||
GenBank mRNA | AB027448.1 | - | 3 | 572 | ||
AB064153.1 | - | 3 | 635 | |||
AB064158.1 | - | 3 | 635 | |||
AB064173.1 | - | 623 | ||||
AB064175.1 | - | 3 | 635 | |||
AB064186.1 | - | 3 | 632 | |||
AB064202.1 | - | 635 | ||||
AB064207.1 | - | 3 | 635 | |||
AB064212.1 | - | 3 | 629 | |||
AB064220.1 | - | 3 | 635 | |||
AB064233.1 | - | 3 | 626 | |||
AJ318021.1 | - | 3 | 840 | |||
AJ318022.1 | - | 3 | 837 | |||
AJ319669.1 | - | 2 | 470 | |||
AJ319670.1 | - | 2 | 669 | |||
AJ319671.1 | - | 2 | 564 | |||
BC012876.2 | - | 4 | 670 | 4149333 | ||
BC062711.1 | - | 4 | 675 | 30350364 | ||
BC064491.1 | - | 4 | 656 | 4576612 | ||
BC073784.1 | - | 4 | 650 | 4297904 | ||
BC073795.1 | - | 4 | 653 | 6302957 | ||
CR936629.1 | - | 4 | 658 | |||
EF589388.1 | - | 3 | 569 | |||
EF589482.1 | - | 3 | 566 | |||
EF589498.1 | - | 3 | 569 | |||
EF589507.1 | - | 3 | 560 | |||
EF589514.1 | - | 3 | 569 | |||
JF806287.1 | - | 4 | 632 | |||
X57819.1 | - | 4 | 627 | |||
X57823.1 | - | 4 | 626 |
Gene Symbol: CYAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB369252.1 | - | 4 | 617 |
Gene Symbol: IGLC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC007782.2 | - | 4 | 635 | ||
BC012159.1 | - | 2 | 659 | 4575311 |
Gene Symbol: IGLJ3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | U07991.1 | - | 3 | 575 | ||
X57812.1 | - | 4 | 629 | |||
X57817.1 | - | 4 | 620 | |||
X57821.1 | - | 4 | 622 |
Gene Symbol: IGLV3-21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC070352.1 | - | 4 | 660 |
Gene Symbol: IGLV3-25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC089418.1 | - | 5 | 632 |
Gene Symbol: IGLV4-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC020236.1 | - | 4 | 673 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
