siRNA Details
Gene Symbol: NPIPB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130464.2 | NP_569731.2 | 9 | 3135 | ||
GenBank mRNA | AB209632.1 | - | 11 | 4844 | ||
AK128772.1 | - | 5 | 3880 | |||
AK294452.1 | - | 5 | 2698 | |||
AK295170.1 | - | 11 | 3260 | |||
AK300277.1 | - | 2830 | ||||
AK303166.1 | - | 3 | 2216 | |||
AK303500.1 | - | 7 | 2755 | |||
AK304162.1 | - | 6 | 3157 | |||
AK316459.1 | - | 7 | 2690 | |||
AY498718.1 | - | 1 | 62 | |||
BC036263.1 | - | 9 | 1770 | 3903371 | ||
BC061522.1 | - | 2 | 952 | 6190707 | ||
BC073943.1 | - | 1 | 743 | |||
BC094882.1 | - | 8 | 1937 | 3093162 | ||
BM710413.1 | - | 1 | 296 | |||
BQ950195.1 | - | 1 | 106 |
Gene Symbol: LOC613037 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321892.1 | NP_001308821.1 | 9 | 3267 | ||
XM_017023514.1 | XP_016879003.1 | 9 | 3556 | |||
XM_017023515.1 | XP_016879004.1 | 10 | 3575 | |||
XM_017023516.1 | XP_016879005.1 | 9 | 3602 | |||
XM_017023517.1 | XP_016879006.1 | 9 | 3333 | |||
XM_017023518.1 | XP_016879007.1 | 9 | 3421 | |||
XM_017023519.1 | XP_016879008.1 | 9 | 3478 | |||
XM_017023520.1 | XP_016879009.1 | 4 | 4542 | |||
XM_017023521.1 | XP_016879010.1 | 5 | 2797 | |||
XM_017023522.1 | XP_016879011.1 | 2 | 3206 |
Gene Symbol: NPIPB11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310137.1 | NP_001297066.1 | 8 | 3113 |
Gene Symbol: NPIPB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310148.1 | NP_001297077.1 | 8 | 3551 | ||
XM_017023244.1 | XP_016878733.1 | 7 | 5562 | |||
XM_017023245.1 | XP_016878734.1 | 8 | 5399 | |||
XM_017023246.1 | XP_016878735.1 | 8 | 5320 | |||
XM_017023247.1 | XP_016878736.1 | 9 | 3307 | |||
GenBank mRNA | AK303607.1 | - | 2573 |
Gene Symbol: NPIPB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135865.1 | NP_001129337.1 | 7 | 3338 | ||
GenBank mRNA | AK293635.1 | - | 1 | 2296 | ||
AK296301.1 | - | 2952 | ||||
AK296338.1 | - | 7 | 3179 | |||
AK303264.1 | - | 6 | 1781 | |||
AK303572.1 | - | 2 | 2468 | |||
AK316332.1 | - | 1 | 1586 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
