siRNA Details
Gene Symbol: COL18A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030582.3 | NP_085059.2 | 39 | 4061 | ||
NM_130444.2 | NP_569711.2 | 39 | 4766 | |||
NM_130445.3 | NP_569712.2 | 40 | 3594 | |||
GenBank mRNA | AF018081.1 | - | 39 | 4080 | ||
AF018082.1 | - | 40 | 3559 | |||
AF184060.1 | - | 2 | 44 | |||
AF416592.1 | - | 2 | 53 | |||
AK098216.1 | - | 6 | 1684 | |||
AK130835.1 | - | 7 | 711 | |||
BC033715.1 | - | 28 | 1942 | |||
BC063833.1 | - | 40 | 3535 | 6181818 | ||
L22548.1 | - | 21 | 1544 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
