siRNA Details
Gene Symbol: USP32P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003190.2 | 6, 7 | 1554 | |||
GenBank mRNA | AK302945.1 | - | 2, 3 | 659 | ||
AK316466.1 | - | 4, 5 | 1272 | |||
AY363305.1 | - | 2, 3 | 936 | |||
BC039345.1 | - | 6, 7 | 1311 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
