siRNA Details
Gene Symbol: TWIST1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000474.3 | NP_000465.1 | 1 | 839 | ||
XM_011515496.1 | XP_011513798.1 | 1 | 586 | |||
GenBank mRNA | BC036704.2 | - | 1 | 839 | 4125830 | |
X99268.1 | - | 1 | 610 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
