siRNA Details
Gene Symbol: RPL15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253379.1 | NP_001240308.1 | 4 | 1027 | ||
NM_001253380.1 | NP_001240309.1 | 3 | 789 | |||
NM_001253382.1 | NP_001240311.1 | 4 | 778 | |||
NM_001253383.1 | NP_001240312.1 | 4 | 757 | |||
NM_002948.3 | NP_002939.2 | 4 | 1014 | |||
GenBank mRNA | AF279903.1 | - | 3 | 668 | ||
AF283772.2 | - | 4 | 674 | |||
AK091751.1 | - | 2 | 1754 | |||
AK098795.1 | - | 4 | 664 | |||
AK129815.1 | - | 4 | 674 | |||
AK290694.1 | - | 4 | 670 | |||
AK292000.1 | - | 4 | 670 | |||
AK293709.1 | - | 4 | 504 | |||
AK297091.1 | - | 5 | 552 | |||
AY347528.1 | - | 3 | 739 | |||
BC014837.1 | - | 4 | 677 | 2901238 | ||
BC068198.1 | - | 1 | 673 | 4093343 | ||
BC070328.1 | - | 4 | 670 | 30371842 | ||
BC071672.1 | - | 1 | 659 | 4390593 | ||
BX537495.1 | - | 2 | 1697 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
