siRNA Details
Gene Symbol: USP17L18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256859.1 | NP_001243788.1 | 1 | 1072 |
Gene Symbol: USP17L10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256852.1 | NP_001243781.1 | 1 | 1072 |
Gene Symbol: USP17L11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256854.1 | NP_001243783.1 | 1 | 1072 |
Gene Symbol: USP17L12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256853.1 | NP_001243782.1 | 1 | 1072 |
Gene Symbol: USP17L15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256894.1 | NP_001243823.1 | 1 | 1072 |
Gene Symbol: USP17L19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256860.1 | NP_001243789.1 | 1 | 1072 |
Gene Symbol: USP17L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY509884.1 | - | 1 | 1072 |
Gene Symbol: USP17L20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256861.1 | NP_001243790.1 | 1 | 1072 |
Gene Symbol: USP17L21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256862.1 | NP_001243791.1 | 1 | 1072 |
Gene Symbol: USP17L22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256863.1 | NP_001243792.1 | 1 | 1072 |
Gene Symbol: USP17L24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242327.1 | NP_001229256.1 | 1 | 1072 |
Gene Symbol: USP17L25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242326.1 | NP_001229255.1 | 1 | 1072 |
Gene Symbol: USP17L26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242328.1 | NP_001229257.1 | 1 | 1072 |
Gene Symbol: USP17L27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242330.1 | NP_001229259.1 | 1 | 1072 |
Gene Symbol: USP17L28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242331.1 | NP_001229260.1 | 1 | 1072 |
Gene Symbol: USP17L29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242332.1 | NP_001229261.1 | 1 | 1072 |
Gene Symbol: USP17L30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256867.1 | NP_001243796.1 | 1 | 1072 |
Gene Symbol: USP17L5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242329.1 | NP_001229258.1 | 1 | 1072 |
Gene Symbol: USP17L6P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027279.1 | 1 | 1072 | |||
GenBank mRNA | AY533200.1 | - | 1 | 1072 |
Gene Symbol: USP17L9P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046416.1 | 1 | 1073 | |||
GenBank mRNA | AF544011.1 | - | 2 | 1072 | ||
BC100991.2 | - | 1 | 1072 | |||
BC100992.2 | - | 1 | 1072 | |||
BC100993.1 | - | 1 | 1075 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
