siRNA Details
Gene Symbol: NACA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113201.2 | NP_001106672.1 | 8 | 1019 | ||
NM_001113202.1 | NP_001106673.1 | 8 | 942 | |||
NM_001113203.2 | NP_001106674.2 | 11 | 3072 | |||
NM_001320193.1 | NP_001307122.1 | 8 | 968 | |||
NM_001320194.1 | NP_001307123.1 | 7 | 1175 | |||
NM_005594.5 | NP_005585.1 | 8 | 993 | |||
XM_006719412.1 | XP_006719475.1 | 8 | 7523 | |||
XM_006719413.2 | XP_006719476.1 | 8 | 6779 | |||
XM_006719414.1 | XP_006719477.1 | 8 | 6521 | |||
XM_006719418.1 | XP_006719481.1 | 7 | 958 | |||
XM_006719420.3 | XP_006719483.1 | 7 | 1214 | |||
XM_011538389.1 | XP_011536691.1 | 9 | 6288 | |||
XM_017019337.1 | XP_016874826.1 | 9 | 3095 | |||
GenBank mRNA | AF054187.1 | - | 8 | 969 | ||
AK090650.1 | - | 8 | 2599 | |||
AK096699.1 | - | 11 | 2829 | |||
AK223085.1 | - | 8 | 699 | |||
AK293209.1 | - | 8 | 561 | |||
AW021139.1 | - | 3 | 565 | |||
AY034001.1 | - | 7 | 663 | |||
AY911673.1 | - | 8 | 685 | |||
BC105120.1 | - | 8 | 860 | |||
BC105122.1 | - | 8 | 860 | |||
BC106041.1 | - | 8 | 983 | |||
X80909.1 | - | 8 | 685 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
