siRNA Details
Gene Symbol: TPSAB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC038416.1 | - | 5 | 1094 | ||
BC051852.1 | - | 5 | 1093 | |||
BC065923.1 | - | 5 | 1299 | 5752632 | ||
M30038.1 | - | 5 | 1101 | |||
M37488.1 | - | 5 | 1089 |
Gene Symbol: TPSB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024164.5 | NP_077078.5 | 6 | 1111 | ||
GenBank mRNA | BC028059.1 | - | 5 | 1100 | 5181815 | |
BC029356.1 | - | 6 | 1111 | 4695458 | ||
M33492.1 | - | 5 | 1086 | |||
M33493.1 | - | 5 | 1065 | |||
S55551.1 | - | 5 | 1095 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
