siRNA Details
Gene Symbol: MATN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003833.4 | NP_003824.2 | 4 | 355 | ||
NM_030590.3 | NP_085080.1 | 3 | 297 | |||
NM_030592.3 | NP_085095.1 | 3 | 267 | |||
XM_005260597.1 | XP_005260654.1 | 3 | 536 | |||
XM_017028113.1 | XP_016883602.1 | 4 | 355 | |||
XM_017028114.1 | XP_016883603.1 | 3 | 536 | |||
XM_017028115.1 | XP_016883604.1 | 3 | 536 | |||
GenBank mRNA | AJ007581.1 | - | 2 | 119 | ||
AK074593.1 | - | 4 | 355 | |||
AK074597.1 | - | 3 | 299 | |||
BC131763.1 | - | 2 | 123 | 40121434 | ||
BC141811.1 | - | 3 | 285 | 40147663 | ||
BC151219.1 | - | 3 | 250 | 40148522 | ||
DA970379.1 | - | 3 | 538 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
