siRNA Details
Gene Symbol: MATN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003833.4 | NP_003824.2 | 9 | 1815 | ||
NM_030590.3 | NP_085080.1 | 7 | 1634 | |||
NM_030592.3 | NP_085095.1 | 6 | 1481 | |||
XM_005260597.1 | XP_005260654.1 | 8 | 1996 | |||
XM_017028113.1 | XP_016883602.1 | 9 | 1815 | |||
XM_017028114.1 | XP_016883603.1 | 7 | 1873 | |||
XM_017028115.1 | XP_016883604.1 | 6 | 1750 | |||
GenBank mRNA | AJ007581.1 | - | 7 | 1579 | ||
AK074495.1 | - | 5 | 1013 | |||
AK074595.1 | - | 7 | 1551 | |||
BC131763.1 | - | 7 | 1583 | 40121434 | ||
BC141811.1 | - | 7 | 1622 | 40147663 | ||
BC151219.1 | - | 8 | 1710 | 40148522 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
