siRNA Details
Gene Symbol: RPS10P7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026667.1 | 1 | 619 | |||
GenBank mRNA | AK055590.1 | - | 1 | 1817 | ||
BC030568.2 | - | 1 | 551 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
