siRNA Details
Gene Symbol: TSPYL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003937.2 | NP_001003937.2 | 1 | 368 | ||
GenBank mRNA | AK097417.1 | - | 1 | 322 | ||
BC068576.1 | - | 1 | 303 | 4374337 | ||
DB073493.1 | - | 1 | 368 |
