siRNA Details
Gene Symbol: OR52L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005173.3 | NP_001005173.3 | 1 | 697 | ||
GenBank mRNA | BC137343.1 | - | 1 | 697 | 9021341 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: OR52L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005173.3 | NP_001005173.3 | 1 | 697 | ||
GenBank mRNA | BC137343.1 | - | 1 | 697 | 9021341 |