siRNA Details
Gene Symbol: FRG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005217.1 | NP_001005217.1 | 4 | 878 | ||
NM_001286820.1 | NP_001273749.1 | 4 | 881 | |||
GenBank mRNA | AY714545.1 | - | 4 | 878 | ||
BC144572.1 | - | 4 | 866 | 9053101 |
Gene Symbol: FRG2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080998.1 | NP_001074467.1 | 4 | 878 | ||
GenBank mRNA | AY744466.1 | - | 4 | 878 |
