siRNA Details
Gene Symbol: OR5AK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005323.1 | NP_001005323.1 | 1 | 13 | ||
GenBank mRNA | BC137198.1 | - | 1 | 55 | 9021195 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: OR5AK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005323.1 | NP_001005323.1 | 1 | 13 | ||
GenBank mRNA | BC137198.1 | - | 1 | 55 | 9021195 |