siRNA Details
Gene Symbol: C3orf18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171740.2 | NP_001165211.1 | 4 | 815 | ||
NM_001171741.2 | NP_001165212.1 | 4 | 610 | |||
NM_001171743.2 | NP_001165214.2 | 4 | 594 | |||
NM_016210.4 | NP_057294.2 | 5 | 904 | |||
XM_011533781.2 | XP_011532083.1 | 3 | 1554 | |||
XM_011533782.2 | XP_011532084.1 | 5 | 892 | |||
XM_011533783.2 | XP_011532085.1 | 4 | 782 | |||
XM_011533784.2 | XP_011532086.1 | 5 | 670 | |||
XM_011533785.2 | XP_011532087.1 | 4 | 608 | |||
XM_011533788.1 | XP_011532090.1 | 3 | 214 | |||
XM_011533790.1 | XP_011532092.1 | 5 | 820 | |||
XM_017006547.1 | XP_016862036.1 | 4 | 755 | |||
GenBank mRNA | AF188706.1 | - | 4 | 575 | ||
AK124438.1 | - | 4 | 655 | |||
AK130227.1 | - | 2 | 311 | |||
AK308074.1 | - | 4 | 478 | |||
AK309486.1 | - | 3 | 374 | |||
AK313128.1 | - | 5 | 778 | |||
AL833938.1 | - | 5 | 863 | |||
BC034766.1 | - | 4 | 688 | 5177784 | ||
DA504094.1 | - | 3 | 393 | |||
DA535127.1 | - | 3 | 250 | |||
DB027651.1 | - | 3 | 214 |
