siRNA Details
Gene Symbol: ZNF771 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142305.1 | NP_001135777.1 | 3 | 1345 | ||
NM_016643.3 | NP_057727.2 | 3 | 1358 | |||
GenBank mRNA | AF242768.1 | - | 2 | 987 | ||
BC011870.2 | - | 2 | 1107 | 2960099 | ||
BC026192.2 | - | 3 | 1144 | 4801104 |
