siRNA Details
Gene Symbol: KLHL7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031710.2 | NP_001026880.2 | 3 | 553 | ||
NM_001172428.1 | NP_001165899.1 | 3 | 553 | |||
NM_018846.4 | NP_061334.4 | 3 | 637 | |||
NR_033328.1 | 4 | 684 | ||||
NR_033329.1 | 4 | 684 | ||||
XM_006715753.2 | XP_006715816.1 | 3 | 548 | |||
XM_006715754.2 | XP_006715817.1 | 4 | 631 | |||
XM_006715755.2 | XP_006715818.1 | 4 | 850 | |||
XM_006715756.2 | XP_006715819.1 | 3 | 719 | |||
XM_006715757.3 | XP_006715820.1 | 3 | 548 | |||
XM_017012439.1 | XP_016867928.1 | 4 | 850 | |||
XM_017012440.1 | XP_016867929.1 | 3 | 548 | |||
XM_017012441.1 | XP_016867930.1 | 4 | 631 | |||
GenBank mRNA | AF111113.1 | - | 3 | 437 | ||
AK056390.1 | - | 4 | 635 | |||
AK290479.1 | - | 3 | 347 | |||
AK297369.1 | - | 478 | ||||
AK297595.1 | - | 2 | 394 | |||
AK299006.1 | - | 3 | 279 | |||
AK302995.1 | - | 4 | 407 | |||
AL136597.1 | - | 3 | 499 | |||
AL834193.1 | - | 1 | 29 | |||
AY028802.1 | - | 3 | 202 | |||
BC009555.1 | - | 4 | 600 | |||
BG704835.1 | - | 3 | 642 | |||
EF560731.1 | - | 4 | 403 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
