siRNA Details
Gene Symbol: RNF138 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191324.1 | NP_001178253.2 | 3 | 506 | ||
NM_016271.4 | NP_057355.2 | 4 | 793 | |||
XM_005258285.1 | XP_005258342.1 | 3 | 570 | |||
GenBank mRNA | AF162680.3 | - | 4 | 556 | ||
AK315761.1 | - | 4 | 683 | |||
AL133557.1 | - | 2 | 149 | |||
BC018107.1 | - | 4 | 434 | 3914953 | ||
BM564125.1 | - | 4 | 742 | |||
BT006878.1 | - | 3 | 335 | |||
CR457150.1 | - | 3 | 335 | |||
DA752192.1 | - | 3 | 447 | |||
DB091985.1 | - | 3 | 506 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
