siRNA Details
Gene Symbol: CNOT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001265612.1 | NP_001252541.1 | 44, 45 | 6779 | ||
NM_016284.4 | NP_057368.3 | 44, 45 | 6794 | |||
GenBank mRNA | AB023224.2 | - | 31, 32 | 4839 | ||
AF110778.1 | - | 4, 5 | 571 | |||
AK056927.1 | - | 21, 22 | 3124 | |||
AK075125.1 | - | 14, 15 | 2093 | |||
AK092885.1 | - | 4, 5 | 1338 | |||
AK301092.1 | - | 4, 5 | 558 | |||
AK302495.1 | - | 21, 22 | 3164 | |||
AL117492.1 | - | 16, 17 | 2633 | |||
AL832769.1 | - | 44, 45 | 6743 | |||
AL833549.1 | - | 44, 45 | 6753 | |||
BC000779.2 | - | 6, 7 | 952 | |||
BC024317.1 | - | 27, 28 | 4193 | |||
BC027969.2 | - | 3, 4 | 408 | |||
CR749237.1 | - | 37, 38 | 5734 | |||
EF553522.1 | - | 44, 45 | 6728 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
