siRNA Details
Gene Symbol: LOC494141 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026541.1 | 2 | 471 | |||
NR_026563.1 | 1 | 616 | ||||
NR_026564.1 | 2 | 471 | ||||
GenBank mRNA | AK131059.1 | - | 2 | 449 | ||
BC026079.1 | - | 1 | 623 | |||
BC026082.1 | - | 2 | 468 | |||
BC034967.1 | - | 1 | 623 | |||
BC044592.1 | - | 1 | 616 | |||
BC056262.1 | - | 2 | 472 | |||
DB462574.1 | - | 2 | 472 |
Gene Symbol: SLC25A51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033412.3 | NP_219480.1 | 3 | 659 | ||
GenBank mRNA | BC008500.1 | - | 3 | 420 | 4286914 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
