siRNA Details
Gene Symbol: RNF175 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173662.2 | NP_775933.1 | 7 | 1066 | ||
XM_005262938.3 | XP_005262995.1 | 7 | 1803 | |||
XM_005262939.3 | XP_005262996.1 | 6 | 868 | |||
XM_005262940.4 | XP_005262997.1 | 5 | 1005 | |||
XM_011531879.2 | XP_011530181.1 | 7 | 1011 | |||
XM_011531881.2 | XP_011530183.1 | 7 | 1150 | |||
XM_011531882.2 | XP_011530184.1 | 7 | 1814 | |||
XM_011531883.2 | XP_011530185.1 | 7 | 1031 | |||
XM_017008047.1 | XP_016863536.1 | 5 | 784 | |||
GenBank mRNA | AK091509.1 | - | 6 | 686 | ||
BC034385.1 | - | 7 | 1067 | 5180678 |
